This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00969507_m1
See other LIPG GE Assays ›
Gene Symbol:
LIPG
Gene Name:
lipase G, endothelial type
Gene Aliases:
EDL, EL, PRO719
Chromosome Location:
Chr.18: 49560699 - 49592908 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00969507_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIPG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308006.1 NP_001294935.1 5 - 6 1086 119
NM_006033.3 NP_006024.1 6 - 7 1308 119
XM_005258390.1 XP_005258447.1 6 - 7 2125 119
XM_011526265.2 XP_011524567.1 5 - 6 1116 119
XM_011526267.1 XP_011524569.1 6 - 7 1695 119
XM_017026095.1 XP_016881584.1 6 - 7 1807 119
GenBank mRNA AF118767.1 - 6 - 7 1282 119
AK291799.1 - 6 - 7 1282 119
AK300333.1 - 5 - 6 1057 119
AK315252.1 - 6 - 7 1310 119
BC060825.1 - 6 - 7 1279 30338950 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800007298.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000175.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LIPG

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
response to nutrient
cell proliferation
phospholipid catabolic process
positive regulation of high-density lipoprotein particle clearance
positive regulation of cholesterol transport
high-density lipoprotein particle remodeling
cholesterol homeostasis
reverse cholesterol transport
lipid digestion
regulation of lipoprotein metabolic process
phospholipid homeostasis
lipoprotein lipase activity
phospholipase activity
heparin binding
phosphatidylcholine 1-acylhydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR18002046.hg.1 ENST00000261292.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18002046.hg.1 ENST00000427224.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18002046.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18002046.hg.1 TC18000175.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000579750.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000579750.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000579750.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000579750.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000579750.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000583083.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000583083.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000583083.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000623277.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000623277.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 ENST00000623277.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay PSR1800149952.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay PSR1800149952.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay PSR1800149952.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay PSR1800149952.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay PSR1800149952.hg.1 TC1800007298.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800149952.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay PSR1800149952.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay PSR1800149952.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay PSR1800149952.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000261292.8 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000427224.6 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000577628.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000579750.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000579750.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000579750.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000579750.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000579750.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000580036.5 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000583083.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000583083.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000583083.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000623277.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000623277.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 ENST00000623277.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_001308006.1 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq
Affymetrix Human Clariom D Assay TC1800007298.hg.1 NM_006033.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0