This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00970171_m1
See other ADGRL3 GE Assays ›
Gene Symbol:
ADGRL3
Gene Name:
adhesion G protein-coupled receptor L3
Gene Aliases:
CIRL3, CL3, LEC3, LPHN3
Chromosome Location:
Chr.4: 61201256 - 62078335 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00970171_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322246.1 NP_001309175.1 21 - 22 3963 90
NM_001322402.1 NP_001309331.1 22 - 23 4167 90
NM_015236.5 NP_056051.2 21 - 22 3963 90
XM_011531788.2 XP_011530090.1 22 - 23 3705 90
XM_017007929.1 XP_016863418.1 23 - 24 6478 90
XM_017007930.1 XP_016863419.1 22 - 23 5430 90
XM_017007931.1 XP_016863420.1 22 - 23 3705 90
XM_017007932.1 XP_016863421.1 22 - 23 3705 90
XM_017007934.1 XP_016863423.1 22 - 23 3690 90
XM_017007936.1 XP_016863425.1 21 - 22 3666 90
XM_017007937.1 XP_016863426.1 21 - 22 5226 90
XM_017007940.1 XP_016863429.1 22 - 23 3705 90
XM_017007942.1 XP_016863431.1 21 - 22 3486 90
GenBank mRNA AK294689.1 - 4 - 5 316 90

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRL3

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
synapse assembly
locomotion involved in locomotory behavior
response to cocaine
positive regulation of synapse assembly
cell-cell adhesion
G-protein coupled receptor activity
calcium ion binding
protein binding
carbohydrate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0