This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00970256_g1
See other MEPCE GE Assays ›
Gene Symbol:
MEPCE
Gene Name:
methylphosphate capping enzyme
Gene Aliases:
BCDIN3
Chromosome Location:
Chr.7: 100428790 - 100434126 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00970256_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MEPCE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001194990.1 NP_001181919.1 3 - 4 1541 75
NM_001194991.1 NP_001181920.1 3 - 4 1126 75
NM_001194992.1 NP_001181921.1 3 - 4 991 75
NM_019606.5 NP_062552.2 2 - 3 2274 75
XM_011516410.2 XP_011514712.1 3 - 4 1599 75
GenBank mRNA AK000264.1 - 2 - 3 929 75
AK055964.1 - 3 - 4 1514 75
AK291366.1 - 2 - 3 2069 75
BC000556.2 - 2 - 3 1999 3162468 75
BC016396.1 - 2 - 3 2274 75
BC018935.2 - 2 - 3 1999 4136145 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700008542.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000620.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MEPCE

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
RNA methylation
snRNA metabolic process
negative regulation of chromatin binding
snRNA modification
positive regulation of G1/S transition of mitotic cell cycle
RNA methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07009479.hg.1 TC07000620.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07009479.hg.1 ENST00000310512.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07009479.hg.1 ENST00000414441.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07009479.hg.1 ENST00000425355.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07009479.hg.1 ENST00000471065.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07009479.hg.1 NM_001194990.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07009479.hg.1 NM_001194991.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07009479.hg.1 NM_001194992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07009479.hg.1 NM_019606.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000479201.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000479201.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000479201.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000497759.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000497759.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000497759.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000497759.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 ENST00000497759.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay PSR0700160135.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay PSR0700160135.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700160135.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay PSR0700160135.hg.1 NM_001194991.1 Refseq
Affymetrix Human Clariom D Assay PSR0700160135.hg.1 NM_001194992.1 Refseq
Affymetrix Human Clariom D Assay PSR0700160135.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay PSR0700160135.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay PSR0700160135.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700160135.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay PSR0700160135.hg.1 NM_001194991.1 Refseq
Affymetrix Human Clariom D Assay PSR0700160135.hg.1 NM_001194992.1 Refseq
Affymetrix Human Clariom D Assay PSR0700160135.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay PSR0700160135.hg.1 TC0700008542.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000310512.3 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000414441.5 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000479201.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000479201.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000479201.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000497759.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000497759.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000497759.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000497759.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 ENST00000497759.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194990.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194991.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_001194992.1 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_019606.5 Refseq
Affymetrix Human Clariom D Assay TC0700008542.hg.1 NM_019606.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0