This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00970546_g1
See other NDUFA2 GE Assays ›
Gene Symbol:
NDUFA2
Gene Name:
NADH:ubiquinone oxidoreductase subunit A2
Gene Aliases:
B8, CD14, CIB8
Chromosome Location:
Chr.5: 140645363 - 140647785 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00970546_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001185012.1 NP_001171941.1 1 - 2 302 80
NM_002488.4 NP_002479.1 1 - 2 302 80
GenBank mRNA AF047185.1 - 1 - 2 156 80
AF077029.1 - 1 - 2 173 80
BC003674.1 - 1 - 2 147 4044385 80
BQ054127.1 - 1 - 2 162 80
CR457016.1 - 1 - 2 100 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500012248.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05001862.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDUFA2

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, NADH to ubiquinone
mitochondrial respiratory chain complex I assembly
NADH dehydrogenase (ubiquinone) activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05025583.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05025583.hg.1 NM_001185012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05025583.hg.1 TC05001862.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000510680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000510680.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05025589.hg.1 ENST00000252102.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05025589.hg.1 NM_001185012.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05025589.hg.1 NM_002488.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05025589.hg.1 NR_033697.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05025589.hg.1 TC05001862.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000510680.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000510680.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay TC0500012248.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185110.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500185110.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185110.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185110.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185110.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay PSR0500185110.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185110.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500185110.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185110.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185110.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185110.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay PSR0500185110.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185110.hg.1 TC0500012248.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500185113.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500185113.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185113.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185113.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay PSR0500185113.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185113.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500185113.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185113.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185113.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay PSR0500185113.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185113.hg.1 TC0500012248.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500185108.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500185108.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185108.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185108.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185108.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay PSR0500185108.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185108.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500185108.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185108.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185108.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185108.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay PSR0500185108.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185108.hg.1 TC0500012248.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500185111.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500185111.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185111.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185111.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185111.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay PSR0500185111.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185111.hg.1 ENST00000252102.8 Ensembl
Affymetrix Human Clariom D Assay PSR0500185111.hg.1 ENST00000502960.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185111.hg.1 ENST00000512088.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500185111.hg.1 NM_001185012.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185111.hg.1 NM_002488.4 Refseq
Affymetrix Human Clariom D Assay PSR0500185111.hg.1 NR_033697.1 Refseq
Affymetrix Human Clariom D Assay PSR0500185111.hg.1 TC0500012248.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 115673 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-dcxfx:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0