This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00971405_m1
See other PARD3 GE Assays ›
Gene Symbol:
PARD3
Gene Name:
par-3 family cell polarity regulator
Gene Aliases:
ASIP, Baz, PAR3, PAR3alpha, PARD-3, PARD3A, PPP1R118, SE2-5L16, SE2-5LT1, SE2-5T2
Chromosome Location:
Chr.10: 34109560 - 34815325 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00971405_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PARD3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001184785.1 NP_001171714.1 18 - 19 2933 134
NM_001184786.1 NP_001171715.1 17 - 18 2894 134
NM_001184787.1 NP_001171716.1 18 - 19 2942 134
NM_001184792.1 NP_001171721.1 18 - 19 2933 134
NM_019619.3 NP_062565.2 18 - 19 2942 134
XM_005252528.4 XP_005252585.1 18 - 19 2921 131
XM_005252532.4 XP_005252589.1 17 - 18 2578 134
XM_005252534.4 XP_005252591.1 18 - 19 2921 131
XM_005252535.4 XP_005252592.1 17 - 18 2882 131
XM_011519569.2 XP_011517871.1 18 - 19 2930 131
XM_011519570.2 XP_011517872.1 17 - 18 2894 134
XM_011519575.2 XP_011517877.1 17 - 18 2617 134
XM_011519576.2 XP_011517878.1 17 - 18 2588 134
XM_011519577.2 XP_011517879.1 16 - 17 2765 134
XM_011519578.2 XP_011517880.1 16 - 17 2763 131
XM_011519579.2 XP_011517881.1 16 - 17 2486 134
XM_011519581.2 XP_011517883.1 18 - 19 2933 134
XM_011519583.2 XP_011517885.1 18 - 19 2933 134
GenBank mRNA AF196185.1 - 18 - 19 2890 134
AF196186.1 - 18 - 19 2890 134
AF454057.1 - 16 - 17 2310 134
AF454058.1 - 16 - 17 2782 131
AF454059.1 - 13 - 14 1894 131
AF467002.1 - 18 - 19 2612 134
AF467003.1 - 18 - 19 2603 134
AF467004.1 - 18 - 19 2612 134
AK024668.1 - 5 - 6 585 134
AK025892.1 - 9 - 10 1380 134
BC071566.1 - 17 - 18 2841 30344732 134

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000010292.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001170.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PARD3

Gene Ontology Categories:

Function(s) Process(es)

protein complex assembly
protein targeting to membrane
cell cycle
establishment or maintenance of cell polarity
transforming growth factor beta receptor signaling pathway
protein kinase C-activating G-protein coupled receptor signaling pathway
axonogenesis
asymmetric cell division
negative regulation of peptidyl-threonine phosphorylation
myelination in peripheral nervous system
positive regulation of myelination
regulation of cellular localization
bicellular tight junction assembly
establishment of epithelial cell polarity
protein binding
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-3,4,5-trisphosphate binding
protein phosphatase binding
phosphatidylinositol-3-phosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 ENST00000340077.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 ENST00000346874.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 ENST00000350537.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 ENST00000374773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 ENST00000374776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 ENST00000374788.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 ENST00000374789.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 ENST00000374790.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 ENST00000374794.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 ENST00000466092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 ENST00000544292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 ENST00000545260.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 ENST00000545693.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 NM_001184785.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 NM_001184786.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 NM_001184787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 NM_001184788.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 NM_001184789.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 NM_001184790.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 NM_001184791.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 NM_001184792.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 NM_001184793.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 NM_001184794.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 NM_019619.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10014766.hg.1 TC10001170.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000340077.9 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000346874.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000350537.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000374773.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000374776.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000374788.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000374789.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000374790.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000374794.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000466092.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000544292.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000545260.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000545693.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184785.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184786.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184787.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184788.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184789.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184790.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184791.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184792.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184793.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184794.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_019619.3 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000340077.9 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000346874.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000350537.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000374773.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000374776.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000374788.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000374789.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000374790.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000374794.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000466092.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000544292.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000545260.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 ENST00000545693.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184785.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184786.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184787.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184788.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184789.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184790.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184791.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184792.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184793.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_001184794.1 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 NM_019619.3 Refseq
Affymetrix Human Clariom D Assay PSR1000170140.hg.1 TC1000010292.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0