This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00971500_m1
See other TMEM155 GE Assays ›
Gene Symbol:
TMEM155
Gene Name:
transmembrane protein 155
Gene Aliases:
-
Chromosome Location:
Chr.4: 121758930 - 121765400 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00971500_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM155
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317841.1 NP_001304770.1 3 - 4 550 141
NM_001317842.1 NP_001304771.1 3 - 4 394 141
GenBank mRNA AK310583.1 - 3 - 4 269 141
BX359599.2 - 3 - 4 422 141
DB116307.1 - 3 - 4 269 141

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001317841.1, NM_001317842.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0