This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00972377_g1
See other PGGHG GE Assays ›
Gene Symbol:
PGGHG
Gene Name:
protein-glucosylgalactosylhydroxylysine glucosidase
Gene Aliases:
ATHL1
Chromosome Location:
Chr.11: 287760 - 295688 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00972377_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGGHG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017018355.1 XP_016873844.1 1 - 2 182 148

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017018355.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

glycosidase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
hydrolase activity
hydrolase activity, acting on glycosyl bonds

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000525217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000525217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000525217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000525217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000525217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000525217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000533924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000533924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000533924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000525217.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000525217.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000525217.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000525217.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000525217.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000525217.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000533924.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000533924.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000533924.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000534742.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006443.hg.1 ENST00000534742.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0