This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00972711_g1
See other GOT1L1 GE Assays ›
Gene Symbol:
GOT1L1
Gene Name:
glutamic-oxaloacetic transaminase 1-like 1
Gene Aliases:
-
Chromosome Location:
Chr.8: 37933548 - 37940502 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00972711_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GOT1L1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152413.2 NP_689626.2 4 - 5 617 64
GenBank mRNA BC029504.1 - 4 - 5 618 5273324 64
BM559469.1 - 618 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800010142.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08001139.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GOT1L1

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

aspartate biosynthetic process
L-aspartate:2-oxoglutarate aminotransferase activity
pyridoxal phosphate binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR08014614.hg.1 TC08001139.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08014614.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08014614.hg.1 NM_152413.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000518826.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000518826.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000518826.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000518826.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000524298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000524298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000524298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000524298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 ENST00000524298.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000518826.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000518826.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000518826.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000518826.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000524298.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000524298.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000524298.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000524298.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 ENST00000524298.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay TC0800010142.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay PSR0800168948.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay PSR0800168948.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay PSR0800168948.hg.1 ENST00000307599.4 Ensembl
Affymetrix Human Clariom D Assay PSR0800168948.hg.1 NM_152413.2 Refseq
Affymetrix Human Clariom D Assay PSR0800168948.hg.1 TC0800010142.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0