This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00973210_m1
See other ZNF41 GE Assays ›
Gene Symbol:
ZNF41
Gene Name:
zinc finger protein 41
Gene Aliases:
MRX89
Chromosome Location:
Chr.X: 47444691 - 47483671 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00973210_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF41
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001324140.1 NP_001311069.1 1 - 2 179 100
NM_001324142.1 NP_001311071.1 1 - 2 179 100
NM_001324153.1 NP_001311082.1 1 - 2 179 100
XM_017029810.1 XP_016885299.1 1 - 2 581 100
GenBank mRNA BQ053634.1 - 1 - 2 123 100

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001324140.1, NM_001324142.1, NM_001324153.1, XM_017029810.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0