This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00973528_gH
See other AKR1D1 GE Assays ›
Gene Symbol:
AKR1D1
Gene Name:
aldo-keto reductase family 1, member D1
Gene Aliases:
3o5bred, CBAS2, SRD5B1
Chromosome Location:
Chr.7: 138076432 - 138118304 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00973528_gH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AKR1D1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190907.1 NP_001177836.1 5 - 6 668 73
NM_005989.3 NP_005980.1 5 - 6 668 73
GenBank mRNA AK058142.1 - 5 - 6 491 73
AK289425.1 - 5 - 6 608 73
AK298421.1 - 5 - 6 669 73
BC130625.1 - 5 - 6 623 40146656 73
BC130627.1 - 5 - 6 623 40146658 73
Z28339.1 - 5 - 6 650 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700013466.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000858.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AKR1D1

Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

bile acid biosynthetic process
cholesterol catabolic process
digestion
C21-steroid hormone metabolic process
androgen metabolic process
bile acid catabolic process
oxidation-reduction process
aldo-keto reductase (NADP) activity
steroid binding
delta4-3-oxosteroid 5beta-reductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07013452.hg.1 TC07000858.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07013452.hg.1 ENST00000242375.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07013452.hg.1 ENST00000297463.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07013452.hg.1 ENST00000411726.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07013452.hg.1 ENST00000432161.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07013452.hg.1 ENST00000468877.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07013452.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07013452.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07013452.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000438242.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000438242.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000438242.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000438242.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000470851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000470851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 ENST00000470851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay PSR0700198964.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700198964.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700198964.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700198964.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay PSR0700198964.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay PSR0700198964.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay PSR0700198964.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700198964.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700198964.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700198964.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay PSR0700198964.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay PSR0700198964.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay PSR0700198964.hg.1 TC0700013466.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000242375.7 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000411726.6 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000432161.5 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000438242.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000438242.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000438242.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000438242.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000470851.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000470851.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 ENST00000470851.1 Ensembl
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190906.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_001190907.1 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq
Affymetrix Human Clariom D Assay TC0700013466.hg.1 NM_005989.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0