This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00973693_g1
See other PRPF19 GE Assays ›
Gene Symbol:
PRPF19
Gene Name:
pre-mRNA processing factor 19
Gene Aliases:
NMP200, PRP19, PSO4, SNEV, UBOX4, hPSO4
Chromosome Location:
Chr.11: 60890547 - 60906589 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00973693_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRPF19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014502.4 NP_055317.1 9 - 10 930 83
GenBank mRNA AJ131186.1 - 9 - 10 930 83
BC008719.2 - 9 - 10 925 2961655 83
BC018665.2 - 9 - 10 925 2961655 83
BC018698.2 - 9 - 10 925 2961655 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100011010.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11001844.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PRPF19

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
spliceosomal tri-snRNP complex assembly
spliceosomal complex assembly
mRNA splicing, via spliceosome
inner cell mass cell proliferation
transcription-coupled nucleotide-excision repair
double-strand break repair via nonhomologous end joining
lipid biosynthetic process
proteasomal protein catabolic process
cellular protein localization
negative regulation of neuron differentiation
positive regulation of mRNA splicing, via spliceosome
positive regulation of astrocyte differentiation
protein K63-linked ubiquitination
signal transduction involved in DNA damage checkpoint
protein binding
ligase activity
ubiquitin-ubiquitin ligase activity
identical protein binding
ubiquitin protein ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11021193.hg.1 ENST00000227524.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11021193.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11021193.hg.1 TC11001844.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000539180.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000539180.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000539180.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000540473.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000540473.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000540473.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay PSR1100183069.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100183069.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay PSR1100183069.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100183069.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay PSR1100183069.hg.1 TC1100011010.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000227524.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000535326.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000539180.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000539180.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000539180.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000539960.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000540473.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000540473.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000540473.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000541371.5 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 ENST00000546152.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq
Affymetrix Human Clariom D Assay TC1100011010.hg.1 NM_014502.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline