This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00974189_g1
See other CDH23 GE Assays ›
Gene Symbol:
CDH23
Gene Name:
cadherin-related 23
Gene Aliases:
CDHR23, USH1D
Chromosome Location:
Chr.10: 71396934 - 71815947 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00974189_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001171933.1 NP_001165404.1 19 - 20 2980 67
NM_001171934.1 NP_001165405.1 19 - 20 2980 67
NM_022124.5 NP_071407.4 66 - 67 9773 67
XM_006717940.3 XP_006718003.1 67 - 68 9827 67
XM_006717942.3 XP_006718005.1 65 - 66 9761 67
XM_011540039.2 XP_011538341.1 66 - 67 9824 67
XM_011540042.2 XP_011538344.1 68 - 69 9737 67
XM_011540043.2 XP_011538345.1 67 - 68 9827 67
XM_011540044.2 XP_011538346.1 67 - 68 9468 67
XM_011540045.2 XP_011538347.1 67 - 68 9827 67
XM_011540046.2 XP_011538348.1 62 - 63 9262 67
XM_011540047.2 XP_011538349.1 56 - 57 8562 67
XM_011540052.2 XP_011538354.1 39 - 40 6102 67
XM_017016499.1 XP_016871988.1 66 - 67 9773 67
XM_017016500.1 XP_016871989.1 66 - 67 9701 67
XM_017016501.1 XP_016871990.1 46 - 47 7329 67
XM_017016502.1 XP_016871991.1 46 - 47 7517 67
XM_017016503.1 XP_016871992.1 36 - 37 5505 67
XM_017016504.1 XP_016871993.1 30 - 31 5290 67
XM_017016505.1 XP_016871994.1 30 - 31 5321 67
XM_017016506.1 XP_016871995.1 29 - 30 4760 67
XM_017016507.1 XP_016871996.1 23 - 24 3582 67
XM_017016508.1 XP_016871997.1 19 - 20 3035 67
GenBank mRNA AB053445.1 - 18 - 19 2919 67
AF312024.1 - 65 - 66 9383 67
AY010111.2 - 65 - 66 9383 67
AY563165.1 - 19 - 20 2980 67
AY563166.1 - 19 - 20 2980 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000007959.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000442.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CDH23

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

calcium ion transport
homophilic cell adhesion via plasma membrane adhesion molecules
visual perception
sensory perception of sound
locomotory behavior
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
photoreceptor cell maintenance
response to stimulus
sensory perception of light stimulus
equilibrioception
regulation of cytosolic calcium ion concentration
inner ear receptor stereocilium organization
calcium ion binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10005264.hg.1 ENST00000224721.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10005264.hg.1 ENST00000398788.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10005264.hg.1 ENST00000398855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10005264.hg.1 ENST00000398860.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10005264.hg.1 NM_001171933.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10005264.hg.1 NM_001171934.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10005264.hg.1 NM_001171935.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10005264.hg.1 NM_001171936.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10005264.hg.1 NM_022124.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10005264.hg.1 TC10000442.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 ENST00000224721.10 Ensembl
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 ENST00000398788.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 ENST00000475158.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 ENST00000619887.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 ENST00000622827.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 NM_001171933.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 NM_001171934.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 NM_001171935.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 NM_001171936.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 NM_022124.5 Refseq
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 ENST00000224721.10 Ensembl
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 ENST00000398788.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 ENST00000475158.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 ENST00000619887.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 ENST00000622827.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 NM_001171933.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 NM_001171934.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 NM_001171935.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 NM_001171936.1 Refseq
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 NM_022124.5 Refseq
Affymetrix Human Clariom D Assay PSR1000154096.hg.1 TC1000007959.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline