This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00974386_m1
See other ADGRL3 GE Assays ›
Gene Symbol:
ADGRL3
Gene Name:
adhesion G protein-coupled receptor L3
Gene Aliases:
CIRL3, CL3, LEC3, LPHN3
Chromosome Location:
Chr.4: 61201256 - 62078335 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00974386_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011531791.2 XP_011530093.1 20 - 21 3639 69
XM_017007933.1 XP_016863422.1 21 - 22 3678 69
XM_017007935.1 XP_016863424.1 21 - 22 3678 69
XM_017007938.1 XP_016863427.1 20 - 21 3624 69
XM_017007939.1 XP_016863428.1 18 - 19 3420 69
XM_017007941.1 XP_016863430.1 21 - 22 3678 69
GenBank mRNA AB018311.1 - 9 - 10 1424 69
AK299651.1 - 3 - 4 293 69

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
synapse assembly
locomotion involved in locomotory behavior
response to cocaine
positive regulation of synapse assembly
cell-cell adhesion
G-protein coupled receptor activity
calcium ion binding
protein binding
carbohydrate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0