This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00974777_m1
See other FREM1 GE Assays ›
Gene Symbol:
FREM1
Gene Name:
FRAS1 related extracellular matrix 1
Gene Aliases:
BNAR, C9orf143, C9orf145, C9orf154, MOTA, TILRR, TRIGNO2
Chromosome Location:
Chr.9: 14734666 - 14911653 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00974777_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FREM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144966.5 NP_659403.4 13 - 14 2985 99
XM_005251382.3 XP_005251439.1 12 - 13 3233 99
XM_017014316.1 XP_016869805.1 12 - 13 3260 99
XM_017014317.1 XP_016869806.1 13 - 14 2803 99
XM_017014318.1 XP_016869807.1 13 - 14 3558 99
XM_017014319.1 XP_016869808.1 14 - 15 3462 99
XM_017014320.1 XP_016869809.1 13 - 14 3389 99
XM_017014321.1 XP_016869810.1 12 - 13 3052 99
XM_017014322.1 XP_016869811.1 12 - 13 2846 99
XM_017014323.1 XP_016869812.1 12 - 13 2519 99
XM_017014324.1 XP_016869813.1 12 - 13 3260 99
XM_017014325.1 XP_016869814.1 12 - 13 3260 99
XM_017014326.1 XP_016869815.1 11 - 12 2076 99
XM_017014327.1 XP_016869816.1 8 - 9 1445 99
XM_017014328.1 XP_016869817.1 12 - 13 3260 99
XM_017014329.1 XP_016869818.1 12 - 13 3260 99
XM_017014330.1 XP_016869819.1 12 - 13 3260 99
GenBank mRNA AB160987.1 - 11 - 12 2169 99
BX537574.1 - 13 - 14 2986 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900009581.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000920.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FREM1

Gene Ontology Categories:

Function(s) Process(es)

cell communication
cell-matrix adhesion
craniofacial suture morphogenesis
carbohydrate binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09012272.hg.1 TC09000920.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09012272.hg.1 ENST00000380880.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012272.hg.1 ENST00000380881.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012272.hg.1 ENST00000422223.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012272.hg.1 NM_144966.5 Refseq
Affymetrix Human Clariom D Assay PSR0900165665.hg.1 ENST00000380875.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900165665.hg.1 ENST00000380880.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900165665.hg.1 ENST00000422223.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900165665.hg.1 NM_144966.5 Refseq
Affymetrix Human Clariom D Assay PSR0900165665.hg.1 ENST00000380875.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900165665.hg.1 ENST00000380880.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900165665.hg.1 ENST00000422223.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900165665.hg.1 NM_144966.5 Refseq
Affymetrix Human Clariom D Assay PSR0900165665.hg.1 TC0900009581.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0