This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00975905_m1
See other IGSF10 GE Assays ›
Gene Symbol:
IGSF10
Gene Name:
immunoglobulin superfamily member 10
Gene Aliases:
CMF608
Chromosome Location:
Chr.3: 151435051 - 151619926 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00975905_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178822.4 NP_849144.2 4 - 5 5062 68
XM_011512708.2 XP_011511010.1 10 - 11 5871 68
XM_011512709.2 XP_011511011.1 9 - 10 5602 68
GenBank mRNA AK128160.1 - 1 - 2 1112 68
AY273815.1 - 4 - 5 5062 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012818.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001908.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGSF10

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s)

ossification
multicellular organism development
cell differentiation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000489791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000489791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000497472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000497472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline