This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00975906_m1
See other IGSF10 GE Assays ›
Gene Symbol:
IGSF10
Gene Name:
immunoglobulin superfamily member 10
Gene Aliases:
CMF608
Chromosome Location:
Chr.3: 151435051 - 151619926 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00975906_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178822.4 NP_849144.2 5 - 6 5964 141
XM_011512708.2 XP_011511010.1 11 - 12 6773 141
XM_011512709.2 XP_011511011.1 10 - 11 6504 141
GenBank mRNA AY273815.1 - 5 - 6 5964 141

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012818.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001908.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGSF10

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s)

ossification
multicellular organism development
cell differentiation

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000282466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000489791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000489791.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000493841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000495443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000497472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 ENST00000497472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178145.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_001178146.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012818.hg.1 NM_178822.4 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 183393 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0