Assay Details
| Gene Symbol: MED12L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053002.5 | NP_443728.3 | 38 - 39 | 5934 | 80 | |
| XM_006713487.3 | XP_006713550.1 | 40 - 41 | 6311 | 80 | ||
| XM_011512390.2 | XP_011510692.1 | 39 - 40 | 6206 | 80 | ||
| XM_017005676.1 | XP_016861165.1 | 41 - 42 | 6362 | 80 | ||
| XM_017005677.1 | XP_016861166.1 | 41 - 42 | 6359 | 80 | ||
| XM_017005679.1 | XP_016861168.1 | 40 - 41 | 6092 | 80 | ||
| XM_017005680.1 | XP_016861169.1 | 39 - 40 | 6080 | 80 | ||
| XM_017005681.1 | XP_016861170.1 | 25 - 26 | 3697 | 80 | ||
| GenBank mRNA | AB046855.1 | - | 25 - 26 | 3677 | 80 | |
| AF399708.1 | - | 38 - 39 | 5843 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

MED12L
Genomic Map