This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00977557_g1
See other IGF2BP1 GE Assays ›
Gene Symbol:
IGF2BP1
Gene Name:
insulin like growth factor 2 mRNA binding protein 1
Gene Aliases:
CRD-BP, CRDBP, IMP-1, IMP1, VICKZ1, ZBP1
Chromosome Location:
Chr.17: 48997412 - 49056145 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00977557_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGF2BP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160423.1 NP_001153895.1 11 - 12 1443 57
NM_006546.3 NP_006537.3 13 - 14 1860 57
XM_005256955.3 XP_005257012.2 12 - 13 1583 57
XM_011524201.2 XP_011522503.1 13 - 14 1808 57
XM_017024022.1 XP_016879511.1 13 - 14 1805 57
GenBank mRNA AF117106.1 - 13 - 14 1535 57
AF198254.1 - 13 - 14 1831 57
AK074915.1 - 8 - 9 863 57
AK313229.1 - 13 - 14 1831 57
DQ227344.1 - 11 - 12 1109 57
KR861348.1 - 12 - 13 1482 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700008186.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000636.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGF2BP1

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

gene expression
regulation of mRNA stability involved in response to stress
negative regulation of translation
pallium cell proliferation in forebrain
regulation of cytokine biosynthetic process
mRNA transport
CRD-mediated mRNA stabilization
neuronal stem cell population maintenance
nucleotide binding
mRNA binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding
translation regulator activity
mRNA 5'-UTR binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 ENST00000578722.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17007962.hg.1 ENST00000290341.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17007962.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17007962.hg.1 NM_001160423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17007962.hg.1 NM_006546.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17007962.hg.1 TC17000636.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1700160567.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700160567.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700160567.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay PSR1700160567.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay PSR1700160567.hg.1 TC1700008186.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700160567.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700160567.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700160567.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay PSR1700160567.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000290341.7 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000431824.2 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000499130.6 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000505562.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000510023.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000515586.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 ENST00000578722.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_001160423.1 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq
Affymetrix Human Clariom D Assay TC1700008186.hg.1 NM_006546.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline