This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00978523_m1
See other IL7 GE Assays ›
Gene Symbol:
IL7
Gene Name:
interleukin 7
Gene Aliases:
IL-7
Chromosome Location:
Chr.8: 78675870 - 78805523 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00978523_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017013397.1 XP_016868886.1 2 - 3 632 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017013397.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

T cell lineage commitment
humoral immune response
cell-cell signaling
positive regulation of cell proliferation
organ morphogenesis
regulation of gene expression
positive regulation of B cell proliferation
negative regulation of apoptotic process
negative regulation of catalytic activity
bone resorption
positive regulation of T cell differentiation
positive regulation of organ growth
homeostasis of number of cells within a tissue
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
cytokine activity
interleukin-7 receptor binding
protein binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0