This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00978574_g1
See other DTL GE Assays ›
Gene Symbol:
DTL
Gene Name:
denticleless E3 ubiquitin protein ligase homolog
Gene Aliases:
CDT2, DCAF2, L2DTL, RAMP
Chromosome Location:
Chr.1: 212035553 - 212105006 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00978574_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DTL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286230.1 NP_001273159.1 8 - 9 1028 83
NM_016448.3 NP_057532.3 9 - 10 1154 83
XM_011509614.1 XP_011507916.1 9 - 10 858 83
GenBank mRNA AF195765.1 - 9 - 10 918 83
AF345896.1 - 9 - 10 939 83
AK000742.1 - 9 - 10 972 83
AK292343.1 - 9 - 10 1026 83
AK303346.1 - 8 - 9 831 83
BC033297.1 - 9 - 10 895 4342989 83
BC033540.1 - 9 - 10 936 4826434 83
DQ641253.1 - 9 - 10 816 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001767.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DTL

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
DNA replication
ubiquitin-dependent protein catabolic process
protein monoubiquitination
cellular response to DNA damage stimulus
response to UV
translesion synthesis
G2 DNA damage checkpoint
DNA damage response, detection of DNA damage
regulation of cell cycle
ubiquitin-protein transferase activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01027694.hg.1 ENST00000366991.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01027694.hg.1 ENST00000475419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01027694.hg.1 ENST00000542077.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01027694.hg.1 NM_016448.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01027694.hg.1 TC01001767.hg.1 HTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline