This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00978688_m1
See other PITPNB GE Assays ›
Gene Symbol:
PITPNB
Gene Name:
phosphatidylinositol transfer protein beta
Gene Aliases:
PI-TP-beta, PtdInsTP, VIB1B
Chromosome Location:
Chr.22: 27851669 - 27919306 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00978688_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PITPNB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284278.1 NP_001271207.1 2 - 3 139 93
XM_011530052.2 XP_011528354.1 2 - 3 174 93
XM_017028707.1 XP_016884196.1 2 - 3 134 93
GenBank mRNA AK302367.1 - 2 - 3 125 93
DC313909.1 - 2 - 3 139 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200008350.hg.1

Set Membership:

Amplicon length between 86 and 100 Transporter Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phospholipid transport
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000460566.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000460566.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000460566.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000460566.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000465179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000465179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000465179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000465179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000471825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000471825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000471825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000493007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000493007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000493007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000493007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline