This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00978692_m1
See other PITPNB GE Assays ›
Gene Symbol:
PITPNB
Gene Name:
phosphatidylinositol transfer protein beta
Gene Aliases:
PI-TP-beta, PtdInsTP, VIB1B
Chromosome Location:
Chr.22: 27851669 - 27919306 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00978692_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PITPNB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284277.1 NP_001271206.1 6 - 7 489 117
NM_001284278.1 NP_001271207.1 6 - 7 467 117
NM_012399.4 NP_036531.1 6 - 7 489 117
XM_011530052.2 XP_011528354.1 6 - 7 502 117
GenBank mRNA AK302367.1 - 6 - 7 453 117
AK313064.1 - 6 - 7 439 117
BC018704.1 - 6 - 7 413 4643509 117
BC031427.1 - 6 - 7 406 4341137 117
BU147254.1 - 6 - 7 450 117
CR456541.1 - 6 - 7 441 117
D30037.1 - 6 - 7 420 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200008350.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000621.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PITPNB

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phospholipid transport
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000460566.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000460566.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000460566.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000460566.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000465179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000465179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000465179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000465179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000471825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000471825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000471825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000493007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000493007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000493007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000493007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline