This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00978693_m1
See other PITPNB GE Assays ›
Gene Symbol:
PITPNB
Gene Name:
phosphatidylinositol transfer protein beta
Gene Aliases:
PI-TP-beta, PtdInsTP, VIB1B
Chromosome Location:
Chr.22: 27851669 - 27919306 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00978693_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PITPNB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284277.1 NP_001271206.1 7 - 8 573 151
NM_001284278.1 NP_001271207.1 7 - 8 551 151
NM_012399.4 NP_036531.1 7 - 8 573 151
XM_011530052.2 XP_011528354.1 7 - 8 586 151
GenBank mRNA AK302367.1 - 7 - 8 537 151
AK313064.1 - 7 - 8 523 151
BC018704.1 - 7 - 8 497 4643509 151
BC031427.1 - 7 - 8 490 4341137 151
BU147254.1 - 7 - 8 534 151
CR456541.1 - 7 - 8 525 151
D30037.1 - 7 - 8 504 151

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200008350.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000621.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PITPNB

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phospholipid transport
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000320996.14 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000335272.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000415296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000436663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000455418.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000460566.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000460566.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000460566.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000460566.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000465179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000465179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000465179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000465179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000471825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000471825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000471825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000477861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000493007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000493007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000493007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000493007.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 ENST00000634017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284277.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_001284278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008350.hg.1 NM_012399.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline