This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00979016_m1
See other APBA1 GE Assays ›
Gene Symbol:
APBA1
Gene Name:
amyloid beta precursor protein binding family A member 1
Gene Aliases:
D9S411E, LIN10, MINT1, X11, X11A, X11ALPHA
Chromosome Location:
Chr.9: 69427532 - 69673012 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00979016_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APBA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163.3 NP_001154.2 6 - 7 1792 102
XM_011518617.2 XP_011516919.1 6 - 7 1747 102
XM_017014670.1 XP_016870159.1 6 - 7 2036 102
GenBank mRNA AF047347.1 - 6 - 7 1755 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900010350.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001193.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene APBA1

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
protein complex assembly
intracellular protein transport
cell adhesion
chemical synaptic transmission
neurotransmitter secretion
nervous system development
locomotory behavior
axo-dendritic transport
regulation of gene expression
glutamate secretion
gamma-aminobutyric acid secretion
multicellular organism growth
beta-amyloid binding
protein binding
phosphatidylinositol-4,5-bisphosphate binding
PDZ domain binding
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000457328.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000457328.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000470082.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000470082.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 ENST00000470082.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09015272.hg.1 TC09001193.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09015272.hg.1 ENST00000265381.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09015272.hg.1 ENST00000470082.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09015272.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000457328.1 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000457328.1 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000470082.2 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000470082.2 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 ENST00000470082.2 Ensembl
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay TC0900010350.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay PSR0900170816.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900170816.hg.1 ENST00000470082.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900170816.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay PSR0900170816.hg.1 ENST00000265381.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900170816.hg.1 ENST00000470082.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900170816.hg.1 NM_001163.3 Refseq
Affymetrix Human Clariom D Assay PSR0900170816.hg.1 TC0900010350.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0