This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00979198_m1
See other GJA5 GE Assays ›
Gene Symbol:
GJA5
Gene Name:
gap junction protein alpha 5
Gene Aliases:
ATFB11, CX40
Chromosome Location:
Chr.1: 147756199 - 147781127 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00979198_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJA5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005266.6 NP_005257.2 1 - 2 106 89
GenBank mRNA BQ231177.1 - 1 - 2 82 89
BQ924948.1 - 1 - 2 98 89
DR003194.1 - 1 - 2 106 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100015636.hg
This assay targets the same location as HTA transcript cluster(s) TC01003129.hg

Additional Information:

This assay is in the 5'UTR region of the NM_005266.6 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GJA5

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
angiogenesis
negative regulation of glomerular filtration
outflow tract morphogenesis
endothelium development
cardiac conduction system development
mitral valve development
pulmonary valve formation
ventricular septum development
atrial septum development
septum primum development
atrial ventricular junction remodeling
potassium ion transport
cell communication by chemical coupling
positive regulation of cell communication by chemical coupling
gap junction assembly
embryonic limb morphogenesis
embryonic heart tube development
foramen ovale closure
negative regulation of blood pressure
positive regulation of vasoconstriction
positive regulation of vasodilation
artery morphogenesis
protein oligomerization
transmembrane transport
regulation of cardiac muscle contraction
regulation of ventricular cardiac muscle cell membrane repolarization
regulation of atrial cardiac muscle cell membrane depolarization
regulation of ventricular cardiac muscle cell membrane depolarization
ventricular septum morphogenesis
atrial septum morphogenesis
ventricular cardiac muscle cell action potential
SA node cell action potential
AV node cell to bundle of His cell communication by electrical coupling
bundle of His cell to Purkinje myocyte communication by electrical coupling
cell communication by electrical coupling involved in cardiac conduction
regulation of heart rate by cardiac conduction
regulation of AV node cell action potential
regulation of bundle of His cell action potential
regulation of Purkinje myocyte action potential
regulation of atrial cardiac muscle cell action potential
regulation of renin secretion into blood stream
regulation of membrane depolarization during cardiac muscle cell action potential
vasomotion
gap junction hemi-channel activity
connexin binding
gap junction channel activity involved in cardiac conduction electrical coupling
gap junction channel activity involved in AV node cell-bundle of His cell electrical coupling

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000430508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000430508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000430508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 NM_005266.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 NM_005266.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 NM_005266.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 NM_005266.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 NM_005266.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 NM_005266.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01048509.hg.1 ENST00000271348.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01048509.hg.1 NM_005266.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01048509.hg.1 TC01003129.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01048508.hg.1 ENST00000271348.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01048508.hg.1 NM_005266.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01048508.hg.1 TC01003129.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100217086.hg.1 ENST00000430508.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217086.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217086.hg.1 NM_005266.6 Refseq
Affymetrix Human Clariom D Assay PSR0100217086.hg.1 ENST00000430508.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217086.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217086.hg.1 NM_005266.6 Refseq
Affymetrix Human Clariom D Assay PSR0100217086.hg.1 TC0100015636.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000430508.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000430508.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000430508.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015636.hg.1 NM_005266.6 Refseq
Affymetrix Human Clariom D Assay TC0100015636.hg.1 NM_005266.6 Refseq
Affymetrix Human Clariom D Assay TC0100015636.hg.1 NM_005266.6 Refseq
Affymetrix Human Clariom D Assay TC0100015636.hg.1 NM_005266.6 Refseq
Affymetrix Human Clariom D Assay TC0100015636.hg.1 NM_005266.6 Refseq
Affymetrix Human Clariom D Assay TC0100015636.hg.1 NM_005266.6 Refseq
Affymetrix Human Clariom D Assay TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Clariom D Assay TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Clariom D Assay TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Clariom D Assay TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Clariom D Assay TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Clariom D Assay TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Clariom D Assay TC0100015636.hg.1 NM_181703.3 Refseq
Affymetrix Human Clariom D Assay PSR0100217083.hg.1 ENST00000430508.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217083.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100217083.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217083.hg.1 NM_005266.6 Refseq
Affymetrix Human Clariom D Assay PSR0100217083.hg.1 NM_181703.3 Refseq
Affymetrix Human Clariom D Assay PSR0100217083.hg.1 ENST00000430508.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217083.hg.1 ENST00000579774.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100217083.hg.1 ENST00000621517.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217083.hg.1 NM_005266.6 Refseq
Affymetrix Human Clariom D Assay PSR0100217083.hg.1 NM_181703.3 Refseq
Affymetrix Human Clariom D Assay PSR0100217083.hg.1 TC0100015636.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline