This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00979298_g1
See other CDH15 GE Assays ›
Gene Symbol:
CDH15
Gene Name:
cadherin 15
Gene Aliases:
CDH14, CDH3, CDHM, MCAD, MRD3
Chromosome Location:
Chr.16: 89171755 - 89195492 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00979298_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004933.2 NP_004924.1 9 - 10 1452 95
GenBank mRNA BC008951.2 - 9 - 10 1427 3027886 95
D83542.1 - 9 - 10 1452 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600008868.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000692.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CDH15

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
adherens junction organization
positive regulation of muscle cell differentiation
calcium ion binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000521087.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000521087.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000521087.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000521087.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000521087.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16009767.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16009767.hg.1 NM_004933.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16009767.hg.1 TC16000692.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1600166163.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600166163.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay PSR1600166163.hg.1 TC1600008868.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600166163.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600166163.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000289746.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000521087.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000521087.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000521087.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000521087.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000521087.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 ENST00000524089.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq
Affymetrix Human Clariom D Assay TC1600008868.hg.1 NM_004933.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline