This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00979819_m1
See other IL17RE GE Assays ›
Gene Symbol:
IL17RE
Gene Name:
interleukin 17 receptor E
Gene Aliases:
-
Chromosome Location:
Chr.3: 9902086 - 9916402 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00979819_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL17RE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001193380.1 NP_001180309.1 3 - 4 369 133
NM_153480.1 NP_705613.1 3 - 4 369 133
NM_153483.2 NP_705616.2 4 - 5 388 133
NR_104198.1 2 - 3 353 133
XM_006712976.1 XP_006713039.1 2 - 3 333 133
XM_011533362.2 XP_011531664.1 3 - 4 304 133
XM_011533363.2 XP_011531665.1 3 - 4 304 133
XM_011533364.2 XP_011531666.1 3 - 4 304 133
GenBank mRNA AF458069.1 - 3 - 4 369 133
AK074238.1 - 3 - 4 349 133
AK096793.1 - 2 - 3 331 133
AK297697.1 - 3 - 4 335 133
AK314476.1 - 3 - 4 354 133
AY358451.1 - 3 - 4 369 133
BC035478.1 - 4 - 5 381 133
BC063110.1 - 3 - 4 367 133
BC137077.1 - 4 - 5 331 9021074 133
BC137078.1 - 4 - 5 331 9021075 133

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK096793.1, AK314476.1, XM_006712976.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IL17RE

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
cytokine-mediated signaling pathway
protein binding
interleukin-17 receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03001026.hg.1 TC03000046.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03001026.hg.1 ENST00000295980.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03001026.hg.1 ENST00000383814.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03001026.hg.1 ENST00000421412.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03001026.hg.1 ENST00000454190.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03001026.hg.1 NM_001193380.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03001026.hg.1 NM_153480.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03001026.hg.1 NM_153483.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0