This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00981877_g1
See other PPP1R12B GE Assays ›
Gene Symbol:
PPP1R12B
Gene Name:
protein phosphatase 1 regulatory subunit 12B
Gene Aliases:
MYPT2, PP1bp55
Chromosome Location:
Chr.1: 202348702 - 202588569 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00981877_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R12B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002481.3 NP_002472.2 16 - 17 2484 134
NM_032103.2 NP_115286.1 4 - 5 839 134
NM_032104.2 NP_115287.1 4 - 5 839 134
XM_017001342.1 XP_016856831.1 17 - 18 2667 134
XM_017001343.1 XP_016856832.1 17 - 18 2667 134
XM_017001344.1 XP_016856833.1 17 - 18 2667 134
XM_017001345.1 XP_016856834.1 17 - 18 2667 134
XM_017001346.1 XP_016856835.1 16 - 17 2484 134
XM_017001347.1 XP_016856836.1 16 - 17 2484 134
XM_017001348.1 XP_016856837.1 16 - 17 2484 134
XM_017001349.1 XP_016856838.1 17 - 18 2667 134
XM_017001350.1 XP_016856839.1 17 - 18 2541 134
XM_017001352.1 XP_016856841.1 7 - 8 934 134
XM_017001353.1 XP_016856842.1 7 - 8 924 134
XM_017001354.1 XP_016856843.1 4 - 5 2318 134
GenBank mRNA AB003062.1 - 16 - 17 2497 134
AB050641.1 - 4 - 5 839 134
AB050642.1 - 4 - 5 839 134
AF324888.1 - 16 - 17 2478 134
BC144699.1 - 16 - 17 2476 9053233 134

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100011219.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001675.hg.1

Additional Information:

This assay is in the 5'UTR region of the AB050641.1,AB050642.1,NM_032103.2,NM_032104.2,XM_017001352.1,XM_017001353.1,XM_017001354.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP1R12B

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
regulation of muscle contraction
signal transduction
positive regulation of catalytic activity
protein binding
enzyme activator activity
phosphatase regulator activity
protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01026015.hg.1 ENST00000290419.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01026015.hg.1 ENST00000336894.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01026015.hg.1 ENST00000367270.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01026015.hg.1 ENST00000391959.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01026015.hg.1 ENST00000406302.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01026015.hg.1 NM_002481.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01026015.hg.1 NM_032103.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01026015.hg.1 NM_032104.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01026015.hg.1 TC01001675.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 ENST00000290419.9 Ensembl
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 ENST00000491336.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 ENST00000498070.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 ENST00000608999.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 NM_002481.3 Refseq
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 NM_032103.2 Refseq
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 NM_032104.2 Refseq
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 ENST00000290419.9 Ensembl
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 ENST00000491336.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 ENST00000498070.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 ENST00000608999.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 NM_002481.3 Refseq
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 NM_032103.2 Refseq
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 NM_032104.2 Refseq
Affymetrix Human Clariom D Assay PSR0100182373.hg.1 TC0100011219.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline