This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00983080_m1
See other SLC1A4 GE Assays ›
Gene Symbol:
SLC1A4
Gene Name:
solute carrier family 1 member 4
Gene Aliases:
ASCT1, SATT, SPATCCM
Chromosome Location:
Chr.2: 64988445 - 65023865 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00983080_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC1A4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001193493.1 NP_001180422.1 3 - 4 278 97
NM_003038.4 NP_003029.2 3 - 4 912 97
XM_006712079.2 XP_006712142.1 3 - 4 278 97
GenBank mRNA AB026689.1 - 3 - 4 650 97
AK126636.1 - 2 - 3 2102 97
AK223486.1 - 3 - 4 874 97
AK295687.1 - 3 - 4 233 97
AK298472.1 - 4 - 5 823 97
AK313052.1 - 3 - 4 876 97
AK315986.1 - 4 - 5 834 97
BC026216.1 - 3 - 4 813 4420405 97
BC072423.1 - 3 - 4 872 6186177 97
DA218921.1 - 3 - 4 280 97
L14595.1 - 3 - 4 812 97
L19444.1 - 3 - 4 791 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200007821.hg
This assay targets the same location as HTA transcript cluster(s) TC02000374.hg

Additional Information:

This assay is in the 5'UTR region of the AK295687.1,NM_001193493.1,XM_006712079.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC1A4

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transport
glutamine transport
L-alanine transport
L-cystine transport
proline transport
L-serine transport
threonine transport
hydroxyproline transport
synaptic transmission, glutamatergic
proline transmembrane transport
cognition
mitophagy in response to mitochondrial depolarization
chloride transmembrane transport
chloride channel activity
amino acid transmembrane transporter activity
L-alanine transmembrane transporter activity
L-cystine transmembrane transporter activity
L-glutamine transmembrane transporter activity
L-proline transmembrane transporter activity
L-serine transmembrane transporter activity
L-threonine transmembrane transporter activity
symporter activity
L-hydroxyproline transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02005676.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02005676.hg.1 ENST00000448784.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02005676.hg.1 ENST00000493121.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02005676.hg.1 ENST00000531327.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02005676.hg.1 NM_001193493.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02005676.hg.1 NM_003038.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02005676.hg.1 TC02000374.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay TC0200007821.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay PSR0200153976.hg.1 TC0200007821.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 ENST00000234256.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 ENST00000471551.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 ENST00000480594.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 ENST00000493121.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 ENST00000531327.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 NM_001193493.1 Refseq
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 NM_003038.4 Refseq
Affymetrix Human Clariom D Assay PSR0200153975.hg.1 TC0200007821.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline