This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00983821_m1
See other NXPE1 GE Assays ›
Gene Symbol:
NXPE1
Gene Name:
neurexophilin and PC-esterase domain family member 1
Gene Aliases:
FAM55A
Chromosome Location:
Chr.11: 114518946 - 114560244 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00983821_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NXPE1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152315.3 NP_689528.2 4 - 5 918 90
XM_011542594.2 XP_011540896.1 6 - 7 1435 90
XM_011542595.2 XP_011540897.1 7 - 8 1547 90
XM_011542596.2 XP_011540898.1 7 - 8 1788 90
XM_011542597.2 XP_011540899.1 6 - 7 1676 90
XM_011542598.2 XP_011540900.1 5 - 6 1376 90
XM_011542599.2 XP_011540901.1 5 - 6 1617 90
GenBank mRNA BC029049.1 - 4 - 5 881 5186472 90
BX648601.1 - 2 - 3 1281 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100012381.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002313.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NXPE1

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11027659.hg.1 ENST00000251921.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11027659.hg.1 ENST00000424269.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11027659.hg.1 ENST00000536271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11027659.hg.1 NM_152315.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11027659.hg.1 TC11002313.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000539878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000539878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000539878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000539878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000539878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 ENST00000539878.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000534921.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000536312.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000539878.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000539878.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000539878.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000539878.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000539878.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 ENST00000539878.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay TC1100012381.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay PSR1100195083.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100195083.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100195083.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100195083.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay PSR1100195083.hg.1 ENST00000251921.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100195083.hg.1 ENST00000424269.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100195083.hg.1 ENST00000536271.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100195083.hg.1 NM_152315.4 Refseq
Affymetrix Human Clariom D Assay PSR1100195083.hg.1 TC1100012381.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0