This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00984414_g1
See other PTPN3 GE Assays ›
Gene Symbol:
PTPN3
Gene Name:
protein tyrosine phosphatase, non-receptor type 3
Gene Aliases:
PTP-H1, PTPH1
Chromosome Location:
Chr.9: 109375694 - 109538395 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00984414_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPN3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145368.1 NP_001138840.1 19 - 20 2021 125
NM_001145369.1 NP_001138841.1 15 - 16 4205 125
NM_001145370.1 NP_001138842.1 14 - 15 4070 125
NM_001145371.1 NP_001138843.1 10 - 11 1639 125
NM_001145372.1 NP_001138844.1 9 - 10 1504 125
NM_002829.3 NP_002820.3 20 - 21 2156 125
NR_026918.1 7 - 8 926 125
XM_006717197.3 XP_006717260.1 20 - 21 2265 125
XM_006717199.3 XP_006717262.1 20 - 21 2290 125
XM_006717201.3 XP_006717264.1 19 - 20 2097 125
XM_006717202.3 XP_006717265.1 17 - 18 1859 125
XM_006717203.3 XP_006717266.1 17 - 18 1818 125
XM_006717204.3 XP_006717267.1 20 - 21 2192 125
XM_011518888.2 XP_011517190.1 20 - 21 2226 125
XM_011518889.2 XP_011517191.1 20 - 21 2896 125
XM_017014955.1 XP_016870444.1 20 - 21 2248 125
XM_017014956.1 XP_016870445.1 18 - 19 1969 125
XM_017014957.1 XP_016870446.1 19 - 20 2057 125
GenBank mRNA AK295875.1 - 18 - 19 1941 125
AK296388.1 - 11 - 12 1565 125
AK303608.1 - 7 - 8 926 125
AK304434.1 - 19 - 20 2021 125
AK308349.1 - 7 - 8 889 125
AK316066.1 - 19 - 20 2021 125
AK316118.1 - 9 - 10 1430 125
BC033716.1 - 2 - 3 182 125
BC126117.1 - 19 - 20 2066 8991833 125
BC143848.1 - 18 - 19 1931 9052363 125
BX648253.1 - 14 - 15 4071 125
BX648735.1 - 15 - 16 2640 125
CR749204.1 - 8 - 9 1083 125
DQ104439.1 - 19 - 20 2044 125
M64572.1 - 19 - 20 2067 125
S39392.1 - 12 - 13 1467 125

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900011163.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001468.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PTPN3

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
peptidyl-tyrosine dephosphorylation
negative regulation of mitotic cell cycle
negative regulation of membrane protein ectodomain proteolysis
liver regeneration
regulation of membrane depolarization during action potential
regulation of sodium ion transmembrane transporter activity
phosphotyrosine binding
protein tyrosine phosphatase activity
protein binding
cytoskeletal protein binding
sodium channel regulator activity
ATPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 ENST00000262539.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 ENST00000374541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 ENST00000394827.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 ENST00000394831.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 ENST00000412145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 ENST00000446349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 ENST00000497739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 NM_001145368.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 NM_001145369.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 NM_001145370.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 NM_001145371.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 NM_001145372.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 NM_002829.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 NR_026918.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09018148.hg.1 TC09001468.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 ENST00000262539.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 ENST00000374541.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 ENST00000412145.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 ENST00000446349.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 ENST00000497739.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NM_001145368.1 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NM_001145369.1 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NM_001145370.1 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NM_001145371.1 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NM_001145372.1 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NM_002829.3 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NR_026918.1 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 ENST00000262539.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 ENST00000374541.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 ENST00000412145.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 ENST00000446349.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 ENST00000497739.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NM_001145368.1 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NM_001145369.1 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NM_001145370.1 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NM_001145371.1 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NM_001145372.1 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NM_002829.3 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 NR_026918.1 Refseq
Affymetrix Human Clariom D Assay PSR0900175949.hg.1 TC0900011163.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline