This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00984713_m1
See other PCED1A GE Assays ›
Gene Symbol:
PCED1A
Gene Name:
PC-esterase domain containing 1A
Gene Aliases:
C20orf81, FAM113A, bA12M19.1
Chromosome Location:
Chr.20: 2835314 - 2841243 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00984713_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCED1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271168.1 NP_001258097.1 1 - 2 378 76
XM_005260804.2 XP_005260861.1 1 - 2 378 76
GenBank mRNA AK026029.1 - 1 - 2 286 76
BC014247.2 - 1 - 2 279 4641068 76
BE302577.1 - 1 - 2 325 76
DB276752.1 - 1 - 2 378 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000008200.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK026029.1,BC014247.2,NM_001271168.1,XM_005260804.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Gene Ontology Categories:

Process(es)

protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000474714.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000474714.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000474714.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000474714.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000487501.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000487501.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000487501.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000487501.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 ENST00000487501.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000474714.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000474714.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000474714.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000474714.5 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000487501.2 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000487501.2 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000487501.2 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000487501.2 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 ENST00000487501.2 Ensembl
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay TC2000008200.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay PSR2000157801.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay PSR2000157801.hg.1 TC2000008200.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000157801.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay PSR2000157799.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000157799.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000157799.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay PSR2000157799.hg.1 TC2000008200.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000157799.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000157799.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000157799.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay PSR2000157800.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000157800.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000157800.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay PSR2000157800.hg.1 TC2000008200.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000157800.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000157800.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000157800.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay PSR2000157791.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000157791.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000157791.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000157791.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000157791.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay PSR2000157791.hg.1 NM_022760.4 Refseq
Affymetrix Human Clariom D Assay PSR2000157791.hg.1 TC2000008200.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000157791.hg.1 ENST00000356872.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000157791.hg.1 ENST00000360652.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000157791.hg.1 ENST00000439542.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000157791.hg.1 ENST00000448755.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000157791.hg.1 NM_001271168.1 Refseq
Affymetrix Human Clariom D Assay PSR2000157791.hg.1 NM_022760.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0