This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00985078_m1
See other ASH2L GE Assays ›
Gene Symbol:
ASH2L
Gene Name:
ASH2 like histone lysine methyltransferase complex subunit
Gene Aliases:
ASH2, ASH2L1, ASH2L2, Bre2
Chromosome Location:
Chr.8: 38104965 - 38140080 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00985078_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASH2L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261832.1 NP_001248761.1 13 - 14 1777 70
XM_006716412.1 XP_006716475.1 14 - 15 2222 70
XM_006716413.2 XP_006716476.1 13 - 14 1642 70
GenBank mRNA AF056717.1 - 13 - 14 1631 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800007321.hg.1

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
positive regulation of cell proliferation
hemopoiesis
response to estrogen
positive regulation of transcription from RNA polymerase II promoter
histone H3-K4 methylation
beta-catenin-TCF complex assembly
DNA binding
protein binding
beta-catenin binding
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K4 specific)
transcription regulatory region DNA binding
metal ion binding
euchromatin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000343823.10 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000428278.6 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000517496.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000520079.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000521652.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000521808.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000524247.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000545394.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 NM_001105214.2 Refseq
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 NM_001261832.1 Refseq
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 NM_001282272.1 Refseq
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 NM_004674.4 Refseq
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000343823.10 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000428278.6 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000517496.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000520079.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000521652.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000521808.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000524247.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 ENST00000545394.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 NM_001105214.2 Refseq
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 NM_001261832.1 Refseq
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 NM_001282272.1 Refseq
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 NM_004674.4 Refseq
Affymetrix Human Clariom D Assay PSR0800150312.hg.1 TC0800007321.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0