This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00986480_g1
See other PTPN5 GE Assays ›
Gene Symbol:
PTPN5
Gene Name:
protein tyrosine phosphatase, non-receptor type 5
Gene Aliases:
PTPSTEP, STEP, STEP61
Chromosome Location:
Chr.11: 18727928 - 18792721 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00986480_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPN5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011520411.2 XP_011518713.1 14 - 15 1973 165
XM_017018440.1 XP_016873929.1 13 - 14 1812 165

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
response to amphetamine
positive regulation of receptor internalization
protein dephosphorylation
positive regulation of neuron projection development
positive regulation of protein import into nucleus, translocation
peptidyl-tyrosine dephosphorylation
exploration behavior
response to immobilization stress
response to drug
negative regulation of MAP kinase activity
positive regulation of long-term synaptic potentiation
positive regulation of neuron death
response to acetylsalicylate
positive regulation of protein targeting to mitochondrion
positive regulation of response to drug
phosphotyrosine binding
protein tyrosine phosphatase activity
protein binding
glutamate receptor binding
mitogen-activated protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0