This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00986932_g1
See other HSD3B7 GE Assays ›
Gene Symbol:
HSD3B7
Gene Name:
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
Gene Aliases:
CBAS1, PFIC4, SDR11E3
Chromosome Location:
Chr.16: 30985189 - 30989152 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00986932_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD3B7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025193.3 NP_079469.2 5 - 6 640 114
XM_005255601.3 XP_005255658.2 5 - 6 621 114
XM_011545960.2 XP_011544262.1 5 - 6 818 114
XM_011545961.1 XP_011544263.1 5 - 6 824 114
GenBank mRNA AF277719.1 - 4 - 5 552 114
BC004929.1 - 5 - 6 627 3626629 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600007505.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000365.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HSD3B7

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

bile acid biosynthetic process
B cell chemotaxis
oxidation-reduction process
3-beta-hydroxy-delta5-steroid dehydrogenase activity
protein binding
cholest-5-ene-3-beta,7-alpha-diol 3-beta-dehydrogenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000562932.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000562932.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000562932.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000562932.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000562932.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000562932.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000574447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000574447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000574447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000574447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000574447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 ENST00000574447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16005324.hg.1 ENST00000297679.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16005324.hg.1 NM_025193.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16005324.hg.1 TC16000365.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1600154896.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600154896.hg.1 NM_025193.3 Refseq
Affymetrix Human Clariom D Assay PSR1600154896.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600154896.hg.1 NM_025193.3 Refseq
Affymetrix Human Clariom D Assay PSR1600154896.hg.1 TC1600007505.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000262520.10 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000297679.9 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000562932.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000562932.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000562932.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000562932.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000562932.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000562932.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000574447.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000574447.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000574447.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000574447.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000574447.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 ENST00000574447.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142777.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_001142778.1 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_025193.3 Refseq
Affymetrix Human Clariom D Assay TC1600007505.hg.1 NM_025193.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0