This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00987570_m1
See other AGPAT3 GE Assays ›
Gene Symbol:
AGPAT3
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 3
Gene Aliases:
LPAAT-GAMMA1, LPAAT3
Chromosome Location:
Chr.21: 43865160 - 43987594 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00987570_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AGPAT3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037553.1 NP_001032642.1 7 - 8 951 80
NM_020132.4 NP_064517.1 8 - 9 1071 80
XM_005261160.4 XP_005261217.1 8 - 9 1072 80
XM_006724029.3 XP_006724092.1 7 - 8 1000 80
XM_006724030.3 XP_006724093.1 7 - 8 968 80
XM_006724031.3 XP_006724094.1 7 - 8 971 80
XM_011529660.2 XP_011527962.1 7 - 8 810 80
XM_011529661.2 XP_011527963.1 7 - 8 1125 80
XM_011529662.2 XP_011527964.1 9 - 10 1775 80
XM_011529663.2 XP_011527965.1 7 - 8 984 80
XM_011529664.2 XP_011527966.1 7 - 8 925 80
XM_011529665.2 XP_011527967.1 9 - 10 1426 80
XM_017028408.1 XP_016883897.1 7 - 8 954 80
XM_017028409.1 XP_016883898.1 7 - 8 1032 80
XM_017028410.1 XP_016883899.1 7 - 8 1037 80
XM_017028411.1 XP_016883900.1 7 - 8 910 80
XM_017028412.1 XP_016883901.1 8 - 9 2003 80
XM_017028413.1 XP_016883902.1 8 - 9 971 80
GenBank mRNA AB040138.1 - 7 - 8 1026 80
AF156774.1 - 8 - 9 1032 80
AF156775.1 - 7 - 8 895 80
AK125804.1 - 5 - 6 3688 80
AY358704.1 - 1120 80
BC011971.1 - 8 - 9 1045 4148915 80
BC040603.1 - 9 - 10 1775 5270905 80
BC063552.1 - 7 - 8 1341 4800514 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100007331.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21000215.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AGPAT3

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

phosphatidic acid biosynthetic process
phospholipid biosynthetic process
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR21002410.hg.1 ENST00000291572.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21002410.hg.1 ENST00000327505.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21002410.hg.1 ENST00000398058.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21002410.hg.1 ENST00000398061.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21002410.hg.1 ENST00000398063.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21002410.hg.1 ENST00000479117.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21002410.hg.1 ENST00000546158.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21002410.hg.1 NM_001037553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21002410.hg.1 NM_020132.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21002410.hg.1 TC21000215.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000291572.12 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000327505.6 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000398058.5 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000398061.5 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000398063.6 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000467358.5 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000479117.5 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000484865.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000546158.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 NM_001037553.1 Refseq
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 NM_020132.4 Refseq
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 TC2100007331.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000291572.12 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000327505.6 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000398058.5 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000398061.5 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000398063.6 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000467358.5 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000479117.5 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000484865.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 ENST00000546158.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 NM_001037553.1 Refseq
Affymetrix Human Clariom D Assay PSR2100149491.hg.1 NM_020132.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0