This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00987827_m1
See other TDG GE Assays ›
Gene Symbol:
TDG
Gene Name:
thymine DNA glycosylase
Gene Aliases:
hTDG
Chromosome Location:
Chr.12: 103965815 - 103988878 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00987827_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TDG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003211.4 NP_003202.3 6 - 7 923 96
XM_011538714.1 XP_011537016.1 5 - 6 753 96
GenBank mRNA AK303631.1 - 5 - 6 750 96
AK309792.1 - 7 - 8 967 96
AK313234.1 - 6 - 7 866 96
BC019925.1 - 6 - 7 877 96
BC037557.1 - 6 - 7 876 5184772 96

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TDG

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
base-excision repair
base-excision repair, AP site formation
mismatch repair
transcription, DNA-templated
embryo development
covalent chromatin modification
protein sumoylation
negative regulation of protein binding
oxidative DNA demethylation
negative regulation of chromatin binding
regulation of gene expression, epigenetic
depyrimidination
DNA demethylation
RNA polymerase II transcription cofactor activity
damaged DNA binding
double-stranded DNA binding
uracil DNA N-glycosylase activity
protein kinase C binding
protein binding
pyrimidine-specific mismatch base pair DNA N-glycosylase activity
DNA N-glycosylase activity
mismatched DNA binding
SUMO binding
protein homodimerization activity
structure-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000266775.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000392872.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000401157.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000436021.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000436021.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000436021.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000436021.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000436021.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000436021.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000436021.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000536395.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000536395.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000536395.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000537100.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000537100.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000537100.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000537100.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000537100.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000537100.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000537100.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000537100.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000537100.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000540956.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000540956.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000540956.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000540956.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000540956.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000540956.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000540956.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000540956.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000540956.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000540956.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000540956.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000542926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000542926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000542926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000542926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000542926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000542926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000542926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000542926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000542926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000542926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000542926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000542926.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544060.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544060.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544060.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544060.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544060.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544060.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544060.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000544861.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000545698.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000545698.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000545698.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000545698.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 ENST00000545698.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008669.hg.1 NM_003211.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline