This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00987855_m1
See other SDSL GE Assays ›
Gene Symbol:
SDSL
Gene Name:
serine dehydratase like
Gene Aliases:
SDH 2, SDS-RS1, TDH, cSDH
Chromosome Location:
Chr.12: 113422265 - 113438277 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00987855_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDSL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001304993.1 NP_001291922.1 5 - 6 553 94
NM_138432.3 NP_612441.1 6 - 7 675 94
XM_005253831.4 XP_005253888.1 5 - 6 552 94
XM_011537846.2 XP_011536148.1 5 - 6 1088 94
XM_017018763.1 XP_016874252.1 5 - 6 552 94
GenBank mRNA AF134473.1 - 5 - 6 499 94
BC009849.2 - 6 - 7 654 4040570 94
BC091479.1 - 5 - 6 530 30344219 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200008949.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000892.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SDSL

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

cellular amino acid metabolic process
biological_process
molecular_function
L-serine ammonia-lyase activity
L-threonine ammonia-lyase activity
pyridoxal phosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12011842.hg.1 ENST00000345635.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12011842.hg.1 ENST00000403593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12011842.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12011842.hg.1 TC12000892.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000547802.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000547802.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000547802.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000551760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000551760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000551760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay PSR1200166559.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200166559.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200166559.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200166559.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200166559.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay PSR1200166559.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay PSR1200166559.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200166559.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200166559.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200166559.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200166559.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay PSR1200166559.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay PSR1200166559.hg.1 TC1200008949.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000345635.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000403593.8 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000546672.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000547802.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000547802.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000547802.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000551760.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000551760.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000551760.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 ENST00000553248.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_001304993.1 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq
Affymetrix Human Clariom D Assay TC1200008949.hg.1 NM_138432.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0