This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00987997_m1
See other SS18L1 GE Assays ›
Gene Symbol:
SS18L1
Gene Name:
SS18L1, nBAF chromatin remodeling complex subunit
Gene Aliases:
CREST, LP2261
Chromosome Location:
Chr.20: 62143720 - 62182510 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00987997_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SS18L1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_125980.1 12 - 13 1818 64
NR_125981.1 11 - 12 1439 64
GenBank mRNA AY203931.1 - 8 - 9 1126 64
BC034494.2 - 11 - 12 1393 4823298 64
BC068993.1 - 12 - 13 1756 5094932 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
dendrite development
covalent chromatin modification
positive regulation of transcription, DNA-templated
positive regulation of dendrite morphogenesis
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline