This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00990135_m1
See other PRTFDC1 GE Assays ›
Gene Symbol:
PRTFDC1
Gene Name:
phosphoribosyl transferase domain containing 1
Gene Aliases:
HHGP
Chromosome Location:
Chr.10: 24848607 - 24952644 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00990135_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRTFDC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282786.1 NP_001269715.1 5 - 6 495 63
NM_020200.6 NP_064585.1 5 - 6 495 63
GenBank mRNA AB209792.1 - 5 - 6 445 63
AF226056.1 - 5 - 6 443 63
AK021950.1 - 5 - 6 455 63
AK222674.1 - 5 - 6 454 63
AK294130.1 - 5 - 6 454 63
BC008662.2 - 5 - 6 457 3871911 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000010070.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001110.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PRTFDC1

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

adenine salvage
guanine salvage
GMP salvage
IMP salvage
GMP catabolic process
hypoxanthine metabolic process
nucleotide binding
magnesium ion binding
hypoxanthine phosphoribosyltransferase activity
protein binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10013953.hg.1 ENST00000320152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10013953.hg.1 ENST00000358336.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10013953.hg.1 ENST00000376378.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10013953.hg.1 NM_020200.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10013953.hg.1 TC10001110.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000376376.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000376376.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000376376.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000376376.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000376376.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000464584.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 ENST00000464584.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000376376.3 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000376376.3 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000376376.3 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000376376.3 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000376376.3 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000464584.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 ENST00000464584.2 Ensembl
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_001282786.1 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay TC1000010070.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay PSR1000168606.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Clariom D Assay PSR1000168606.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000168606.hg.1 NM_001282786.1 Refseq
Affymetrix Human Clariom D Assay PSR1000168606.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay PSR1000168606.hg.1 ENST00000320152.10 Ensembl
Affymetrix Human Clariom D Assay PSR1000168606.hg.1 ENST00000376378.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000168606.hg.1 NM_001282786.1 Refseq
Affymetrix Human Clariom D Assay PSR1000168606.hg.1 NM_020200.6 Refseq
Affymetrix Human Clariom D Assay PSR1000168606.hg.1 TC1000010070.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0