This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00990596_m1
See other CD200R1 GE Assays ›
Gene Symbol:
CD200R1
Gene Name:
CD200 receptor 1
Gene Aliases:
CD200R, HCRTR2, MOX2R, OX2R
Chromosome Location:
Chr.3: 112922685 - 112975090 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00990596_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD200R1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138806.3 NP_620161.1 2 - 3 369 124
NM_138939.2 NP_620385.1 2 - 3 369 124
GenBank mRNA AF495380.1 - 2 - 3 191 124
AF497549.1 - 2 - 3 191 124
AK293071.1 - 2 - 3 314 124
AY284975.1 - 2 - 3 358 124
AY358910.1 - 2 - 3 394 124
BC069661.1 - 2 - 3 167 7262527 124
BC069721.1 - 2 - 3 167 7262539 124
BC069743.1 - 2 - 3 167 7262551 124
BC143393.1 - 2 - 3 215 9051899 124

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CD200R1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

viral process
regulation of immune response
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-vvsgg:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0