Assay Details
| Gene Symbol: CD200R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138806.3 | NP_620161.1 | 3 - 4 | 430 | 125 | |
| NM_138939.2 | NP_620385.1 | 3 - 4 | 430 | 125 | ||
| GenBank mRNA | AF495380.1 | - | 3 - 4 | 252 | 125 | |
| AF497549.1 | - | 3 - 4 | 252 | 125 | ||
| AK293071.1 | - | 3 - 4 | 375 | 125 | ||
| AY284975.1 | - | 3 - 4 | 419 | 125 | ||
| AY358910.1 | - | 3 - 4 | 455 | 125 | ||
| BC069661.1 | - | 3 - 4 | 228 | 7262527 | 125 | |
| BC069721.1 | - | 3 - 4 | 228 | 7262539 | 125 | |
| BC069743.1 | - | 3 - 4 | 228 | 7262551 | 125 | |
| BC143393.1 | - | 3 - 4 | 276 | 9051899 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03029819.hg.1 | ENST00000440122.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029819.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029819.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029819.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029819.hg.1 | TC03001654.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000295863.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000308611.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000440122.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000471858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | ENST00000490004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138939.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_138940.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_170780.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012007.hg.1 | NM_170780.2 | Refseq |

CD200R1
Genomic Map