This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00990600_g1
See other CD200R1 GE Assays ›
Gene Symbol:
CD200R1
Gene Name:
CD200 receptor 1
Gene Aliases:
CD200R, HCRTR2, MOX2R, OX2R
Chromosome Location:
Chr.3: 112922685 - 112975090 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00990600_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD200R1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138806.3 NP_620161.1 6 - 7 1108 98
NM_170780.2 NP_740750.1 5 - 6 1039 98
GenBank mRNA AF283760.1 - 5 - 6 814 98
AF495380.1 - 6 - 7 930 98
AF497548.1 - 5 - 6 861 98
AK126349.1 - 5 - 6 1039 98
AK293071.1 - 6 - 7 1053 98
AY284975.1 - 6 - 7 1097 98
AY358910.1 - 6 - 7 1133 98
BC069661.1 - 6 - 7 906 7262527 98
BC069721.1 - 6 - 7 906 7262539 98
BC069743.1 - 6 - 7 906 7262551 98
BC093890.1 - 5 - 6 885 7939735 98
BC143393.1 - 6 - 7 954 9051899 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012007.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001654.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CD200R1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

viral process
regulation of immune response
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03029815.hg.1 ENST00000308611.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029815.hg.1 ENST00000471858.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029815.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03029815.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03029815.hg.1 TC03001654.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay PSR0300189396.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300189396.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189396.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay PSR0300189396.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay PSR0300189396.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300189396.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300189396.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay PSR0300189396.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay PSR0300189396.hg.1 TC0300012007.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline