This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00990637_m1
See other HACD4 GE Assays ›
Gene Symbol:
HACD4
Gene Name:
3-hydroxyacyl-CoA dehydratase 4
Gene Aliases:
PTPLAD2
Chromosome Location:
Chr.9: 21003621 - 21031712 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00990637_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HACD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001010915.4 NP_001010915.2 4 - 5 470 132
NM_001321883.1 NP_001308812.1 4 - 5 407 132
NM_001321903.1 NP_001308832.1 4 - 5 470 132
XM_011517876.2 XP_011516178.1 3 - 4 345 132
XM_017014713.1 XP_016870202.1 4 - 5 660 132
XM_017014714.1 XP_016870203.1 4 - 5 465 132
GenBank mRNA BC114215.1 - 4 - 5 404 5174279 132
BX648779.1 - 4 - 5 432 132
DA333010.1 - 4 - 5 407 132
HY118500.1 - 4 - 5 470 132

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900009685.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000945.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_011517876.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HACD4

Gene Ontology Categories:

Function(s) Process(es)

fatty acid elongation
very long-chain fatty acid biosynthetic process
3-hydroxyacyl-CoA dehydratase activity
enzyme binding
3-hydroxy-behenoyl-CoA dehydratase activity
3-hydroxy-lignoceroyl-CoA dehydratase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09012644.hg.1 TC09000945.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09012644.hg.1 ENST00000495827.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012644.hg.1 ENST00000513293.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012644.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay PSR0900166267.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900166267.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay PSR0900166267.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900166267.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay PSR0900166267.hg.1 TC0900009685.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0