This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00992348_m1
See other KMT5B GE Assays ›
Gene Symbol:
KMT5B
Gene Name:
lysine methyltransferase 5B
Gene Aliases:
CGI-85, CGI85, SUV420H1
Chromosome Location:
Chr.11: 68154863 - 68213904 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00992348_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KMT5B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300907.1 NP_001287836.1 7 - 8 1009 88
NM_001300909.1 NP_001287838.1 5 - 6 960 88
NM_016028.4 NP_057112.3 6 - 7 913 88
NM_017635.4 NP_060105.3 6 - 7 941 88
XM_005274035.3 XP_005274092.2 6 - 7 1161 88
XM_005274036.3 XP_005274093.2 5 - 6 1092 88
XM_006718581.1 XP_006718644.1 5 - 6 823 88
XM_011545091.1 XP_011543393.1 5 - 6 735 88
XM_011545092.2 XP_011543394.1 5 - 6 925 88
XM_011545093.2 XP_011543395.1 6 - 7 1161 88
XM_011545094.2 XP_011543396.1 6 - 7 1161 88
XM_017017876.1 XP_016873365.1 7 - 8 1522 88
XM_017017877.1 XP_016873366.1 7 - 8 1308 88
XM_017017878.1 XP_016873367.1 7 - 8 1253 88
XM_017017879.1 XP_016873368.1 7 - 8 1054 88
XM_017017880.1 XP_016873369.1 6 - 7 1161 88
GenBank mRNA AF151843.1 - 5 - 6 655 88
AK025183.1 - 5 - 6 601 88
AK025461.1 - 1 - 2 65 88
AK299201.1 - 5 - 6 789 88
BC002522.2 - 6 - 7 860 88
BC065287.1 - 6 - 7 865 88
BC087834.1 - 6 - 7 882 88
BC098121.1 - 5 - 6 662 40001705 88
BC099714.1 - 5 - 6 662 40001707 88
BC103498.1 - 5 - 6 662 40001706 88
BC104483.1 - 5 - 6 662 40001704 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC098121.1, BC104483.1, XM_011545093.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene KMT5B

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
muscle organ development
histone H4-K20 trimethylation
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H4-K20 specific)

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0