This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00992730_m1
See other MGST2 GE Assays ›
Gene Symbol:
MGST2
Gene Name:
microsomal glutathione S-transferase 2
Gene Aliases:
GST2, MGST-II
Chromosome Location:
Chr.4: 139665725 - 139754546 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00992730_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MGST2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011531972.2 XP_011530274.1 1 - 2 79 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011531972.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glutathione biosynthetic process
xenobiotic metabolic process
response to organonitrogen compound
leukotriene biosynthetic process
response to lipopolysaccharide
positive regulation of catalytic activity
oxidation-reduction process
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
leukotriene-C4 synthase activity
glutathione peroxidase activity
enzyme activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000502587.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000502587.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000502587.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515067.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000265498.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000502587.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000502587.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000502587.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000503816.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000506797.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515067.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515067.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515067.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515067.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515067.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515067.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000515137.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 ENST00000616265.4 Ensembl
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204366.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204367.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_001204368.1 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_002413.4 Refseq
Affymetrix Human Clariom D Assay TC0400008813.hg.1 NM_002413.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0