This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00994200_m1
See other TWSG1 GE Assays ›
Gene Symbol:
TWSG1
Gene Name:
twisted gastrulation BMP signaling modulator 1
Gene Aliases:
TSG
Chromosome Location:
Chr.18: 9334767 - 9402420 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00994200_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TWSG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020648.5 NP_065699.1 3 - 4 413 75
GenBank mRNA AF196834.1 - 3 - 4 370 75
AF294628.1 - 2 - 3 235 75
AJ297391.1 - 3 - 4 364 75
AK075550.1 - 3 - 4 413 75
AK315751.1 - 3 - 4 298 75
BC020490.1 - 3 - 4 327 3889103 75
DA735800.1 - 3 - 4 413 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800006635.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000045.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TWSG1

Gene Ontology Categories:

Function(s) Process(es)

ossification
mesoderm formation
salivary gland morphogenesis
hemopoiesis
cell differentiation
BMP signaling pathway
positive regulation of BMP signaling pathway
negative regulation of BMP signaling pathway
forebrain development
camera-type eye development
negative regulation of osteoblast differentiation
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR18000487.hg.1 ENST00000262120.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18000487.hg.1 NM_020648.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18000487.hg.1 TC18000045.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Clariom D Assay PSR1800145386.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Clariom D Assay PSR1800145386.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145386.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Clariom D Assay PSR1800145386.hg.1 NM_020648.5 Refseq
Affymetrix Human Clariom D Assay PSR1800145386.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Clariom D Assay PSR1800145386.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800145386.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Clariom D Assay PSR1800145386.hg.1 NM_020648.5 Refseq
Affymetrix Human Clariom D Assay PSR1800145386.hg.1 TC1800006635.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000262120.9 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000581641.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 ENST00000583147.5 Ensembl
Affymetrix Human Clariom D Assay TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Clariom D Assay TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Clariom D Assay TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Clariom D Assay TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Clariom D Assay TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Clariom D Assay TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Clariom D Assay TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Clariom D Assay TC1800006635.hg.1 NM_020648.5 Refseq
Affymetrix Human Clariom D Assay TC1800006635.hg.1 NM_020648.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0