This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00995376_g1
See other SSBP1 GE Assays ›
Gene Symbol:
SSBP1
Gene Name:
single stranded DNA binding protein 1
Gene Aliases:
Mt-SSB, SOSS-B1, SSBP, mtSSB
Chromosome Location:
Chr.7: 141738309 - 141750488 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00995376_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SSBP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256510.1 NP_001243439.1 4 - 5 663 80
NM_001256511.1 NP_001243440.1 4 - 5 544 80
NM_001256512.1 NP_001243441.1 4 - 5 526 80
NM_001256513.1 NP_001243442.1 4 - 5 454 80
NM_003143.2 NP_003134.1 4 - 5 361 80
NR_046269.1 5 - 6 411 80
XM_005250048.4 XP_005250105.1 4 - 5 373 80
XM_005250049.4 XP_005250106.1 4 - 5 412 80
XM_005250050.4 XP_005250107.1 4 - 5 463 80
XM_005250051.4 XP_005250108.1 4 - 5 489 80
GenBank mRNA AF277319.1 - 4 - 5 307 80
AK294393.1 - 4 - 5 317 80
AK313033.1 - 4 - 5 374 80
BC000895.1 - 4 - 5 663 3464282 80
BC093054.1 - 4 - 5 323 30722138 80
BF131408.1 - 5 - 6 341 80
BG030060.1 - 477 80
BP308841.1 - 3 - 4 230 80
BQ959702.1 - 4 - 5 426 80
BU532904.1 - 4 - 5 364 80
M94556.1 - 4 - 5 306 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700013469.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000892.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SSBP1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
mitochondrion organization
positive regulation of helicase activity
mitochondrion morphogenesis
chromatin binding
single-stranded DNA binding
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000461433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000461433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000461433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000461433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000468267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000468267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000468267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000468267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000473783.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000473783.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000473783.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000473783.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000496622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000496622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000496622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000496622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0