This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00996706_m1
See other MGST2 GE Assays ›
Gene Symbol:
MGST2
Gene Name:
microsomal glutathione S-transferase 2
Gene Aliases:
GST2, MGST-II
Chromosome Location:
Chr.4: 139665725 - 139754546 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00996706_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MGST2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204366.1 NP_001191295.1 2 - 3 410 89
NM_002413.4 NP_002404.1 2 - 3 410 89
XM_017008211.1 XP_016863700.1 2 - 3 422 89
XM_017008212.1 XP_016863701.1 2 - 3 422 89
XM_017008213.1 XP_016863702.1 2 - 3 422 89
GenBank mRNA AA122237.1 - 2 - 3 410 89
AK130948.1 - 2 - 3 359 89
BC025416.1 - 2 - 3 324 3937750 89
CR407640.1 - 2 - 3 158 89
DB257575.1 - 2 - 3 174 89
U77604.1 - 2 - 3 169 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MGST2

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glutathione biosynthetic process
xenobiotic metabolic process
response to organonitrogen compound
leukotriene biosynthetic process
response to lipopolysaccharide
positive regulation of catalytic activity
oxidation-reduction process
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
leukotriene-C4 synthase activity
glutathione peroxidase activity
enzyme activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0