This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00996916_m1
See other PLA2G4A GE Assays ›
Gene Symbol:
PLA2G4A
Gene Name:
phospholipase A2 group IVA
Gene Aliases:
PLA2G4, cPLA2, cPLA2-alpha
Chromosome Location:
Chr.1: 186828900 - 186989896 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00996916_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLA2G4A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001311193.1 NP_001298122.1 3 - 4 316 108
NM_024420.2 NP_077734.1 3 - 4 316 108
XM_011509642.2 XP_011507944.1 3 - 4 267 108
GenBank mRNA AK290336.1 - 3 - 4 267 108
AK302938.1 - 3 - 4 226 108
BC114340.2 - 3 - 4 213 40034995 108
DA451491.1 - 3 - 4 267 108
M68874.1 - 3 - 4 249 108
M72393.1 - 3 - 4 236 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100010939.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001607.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PLA2G4A

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

ovulation from ovarian follicle
luteolysis
phospholipid metabolic process
phosphatidic acid biosynthetic process
platelet activating factor biosynthetic process
icosanoid metabolic process
aging
positive regulation of cell proliferation
phospholipid catabolic process
response to heat
response to lithium ion
response to organonitrogen compound
arachidonic acid metabolic process
positive regulation of bone mineralization
positive regulation of vesicle fusion
positive regulation of prostaglandin biosynthetic process
positive regulation of fever generation
response to lipopolysaccharide
response to vitamin D
cardiolipin acyl-chain remodeling
phosphatidylglycerol acyl-chain remodeling
phosphatidylinositol acyl-chain remodeling
phosphatidylserine acyl-chain remodeling
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
response to hydrogen peroxide
positive regulation of apoptotic process
surfactant homeostasis
icosanoid biosynthetic process
decidualization
arachidonic acid secretion
response to glucocorticoid
response to calcium ion
response to methylmercury
cellular response to antibiotic
lysophospholipase activity
phospholipase A2 activity
calcium ion binding
calcium-dependent phospholipid binding
phosphatidylcholine 1-acylhydrolase activity
histone acetyltransferase binding
calcium-dependent phospholipase A2 activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000367466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 ENST00000466600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_001311193.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010939.hg.1 NM_024420.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0